Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001091779.4 | XP_001091779.1 | 452 | 119 | ||
XM_015148229.2 | XP_015003715.2 | 452 | 119 | |||
XM_015148230.2 | XP_015003716.2 | 565 | 119 | |||
XM_015148231.2 | XP_015003717.2 | 344 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |