Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015110089.2 | XP_014965575.1 | 422 | 118 | ||
GenBank mRNA | JU334414.1 | - | 324 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |