Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001095487.4 | XP_001095487.1 | 630 | 97 | ||
GenBank mRNA | JV650826.1 | - | 112 | 97 |