Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC178 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001101213.4 | XP_001101213.2 | 733 | 88 | ||
XM_015121789.2 | XP_014977275.2 | 733 | 88 | |||
XM_015121790.2 | XP_014977276.2 | 664 | 88 | |||
XM_015121791.2 | XP_014977277.2 | 821 | 88 | |||
XM_015121792.2 | XP_014977278.2 | 673 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |