Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCN1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258167.1 | NP_001245096.1 | 61 | 130 | ||
XM_028838435.1 | XP_028694268.1 | 397 | 130 | |||
XM_028838437.1 | XP_028694270.1 | 396 | 130 | |||
GenBank mRNA | JU321213.1 | - | 173 | 130 | ||
JU473653.1 | - | 61 | 130 | |||
JV667885.1 | - | 175 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |