Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015148734.2 | XP_015004220.2 | 614 | 63 | ||
XM_015148744.2 | XP_015004230.1 | 531 | 63 | |||
XM_028830243.1 | XP_028686076.1 | 2658 | 63 | |||
GenBank mRNA | JU333078.1 | - | 277 | 63 | ||
JV707710.1 | - | 469 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |