Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFCAB12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001094977.4 | XP_001094977.2 | 1403 | 107 | ||
GenBank mRNA | JU495854.1 | - | 1373 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |