Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU475236.1 | - | 237 | 86 | ||
KF877035.1 | - | 173 | 86 | |||
KT378619.1 | - | 297 | 86 |