Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001091855.3 | XP_001091855.2 | 347 | 85 | ||
XM_028844421.1 | XP_028700254.1 | 356 | 85 | |||
GenBank mRNA | JV579236.1 | - | 77 | 85 | ||
JV600273.1 | - | 72 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |