Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4V2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193838.2 | NP_001180767.2 | 1706 | 141 | ||
XM_015139562.2 | XP_014995048.2 | 1328 | 141 | |||
XM_028847981.1 | XP_028703814.1 | 1240 | 141 | |||
GenBank mRNA | JU322982.1 | - | 1446 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |