Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001094024.4 | XP_001094024.1 | 172 | 135 | ||
XM_015114997.1 | XP_014970483.1 | 219 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |