Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSH2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190903.2 | NP_001177832.2 | 220 | 99 | ||
XM_015123192.2 | XP_014978678.2 | 384 | 99 | |||
XM_015123193.2 | XP_014978679.2 | 287 | 99 | |||
GenBank mRNA | JU495189.1 | - | 120 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |