Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR10A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194102.2 | NP_001181031.1 | 553 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |