Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR4K2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193863.3 | NP_001180792.2 | 656 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |