Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC10A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015145616.2 | XP_015001102.1 | 3606 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |