Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5AR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015124344.2 | XP_014979830.1 | 1414 | 97 | ||
GenBank mRNA | JV704606.1 | - | 97 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Recommended Assay Macaca mulatta |