Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001098128.3 | XP_001098128.2 | 756 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |