Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015122587.2 | XP_014978073.1 | 942 | 149 | ||
XM_028838923.1 | XP_028694756.1 | 720 | 149 | |||
GenBank mRNA | JU472821.1 | - | 534 | 149 | ||
JV652581.1 | - | 541 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta Inventoried |