Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAB21L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001082398.4 | XP_001082398.1 | 643 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |