Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YWHAQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015111684.2 | XP_014967170.1 | 2201 | 106 | ||
GenBank mRNA | JU474100.1 | - | 2017 | 106 | ||
JV569571.1 | - | 2103 | 106 | |||
JV640825.1 | - | 2064 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |