Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU338342.1 | - | 535 | 102 | ||
JV704573.1 | - | 425 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |