Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRPRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190876.1 | NP_001177805.1 | 597 | 83 | ||
XM_015132068.2 | XP_014987554.1 | 974 | 83 | |||
XM_028843441.1 | XP_028699274.1 | 1029 | 83 | |||
XM_028843442.1 | XP_028699275.1 | 876 | 83 | |||
GenBank mRNA | CO726033.1 | - | 350 | 83 | ||
JU335721.1 | - | 652 | 83 | |||
JU476609.1 | - | 677 | 83 | |||
JV515487.1 | - | 475 | 83 | |||
JV589438.1 | - | 610 | 83 | |||
JV691972.1 | - | 611 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |