Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSME2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257629.1 | NP_001244558.1 | 146 | 165 | ||
XM_028850499.1 | XP_028706332.1 | 184 | 165 | |||
GenBank mRNA | JU320761.1 | - | 146 | 165 | ||
JU472898.1 | - | 153 | 165 | |||
JV043978.1 | - | 153 | 165 | |||
JV636689.1 | - | 154 | 165 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |