Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YKT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266540.1 | NP_001253469.1 | 391 | 155 | ||
GenBank mRNA | JU320009.1 | - | 429 | 155 | ||
JU474036.1 | - | 403 | 155 | |||
JV045961.1 | - | 391 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |