Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORAI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265868.1 | NP_001252797.1 | 375 | 97 | ||
XM_015134538.2 | XP_014990024.1 | 251 | 97 | |||
XM_028845039.1 | XP_028700872.1 | 426 | 97 | |||
XM_028845040.1 | XP_028700873.1 | 331 | 97 | |||
GenBank mRNA | JU336493.1 | - | 285 | 97 | ||
JU472743.1 | - | 375 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |