Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSM12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266793.1 | NP_001253722.1 | 661 | 146 | ||
GenBank mRNA | JU319719.1 | - | 731 | 146 | ||
JU470638.1 | - | 777 | 146 | |||
JV046169.1 | - | 661 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |