Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAMU-F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042770.2 | NP_001036235.2 | 619 | 153 | ||
GenBank mRNA | DQ367724.1 | - | 619 | 153 | ||
EF362454.1 | - | 619 | 153 | |||
EU109708.1 | - | 619 | 153 | |||
EU305657.1 | - | 619 | 153 | |||
FJ211397.1 | - | 595 | 153 | |||
FJ211405.1 | - | 595 | 153 | |||
HM048899.1 | - | 619 | 153 | |||
LT899414.1 | - | 619 | 153 | |||
LT899415.1 | - | 619 | 153 | |||
LT899416.1 | - | 619 | 153 | |||
LT899417.1 | - | 619 | 153 | |||
LT899418.1 | - | 619 | 153 | |||
LT899419.1 | - | 619 | 153 | |||
LT899420.1 | - | 619 | 153 | |||
LT899421.1 | - | 619 | 153 | |||
LT899422.1 | - | 619 | 153 | |||
LT899423.1 | - | 619 | 153 | |||
LT899424.1 | - | 619 | 153 | |||
LT899425.1 | - | 619 | 153 | |||
LT899426.1 | - | 619 | 153 | |||
LT899427.1 | - | 619 | 153 | |||
LT899428.1 | - | 619 | 153 | |||
LT899429.1 | - | 619 | 153 | |||
LT899430.1 | - | 619 | 153 | |||
LT899431.1 | - | 619 | 153 | |||
LT899432.1 | - | 619 | 153 | |||
LT899433.1 | - | 619 | 153 | |||
LT899434.1 | - | 619 | 153 | |||
LT899435.1 | - | 619 | 153 | |||
Z21819.2 | - | 619 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |