Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMELX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098513.2 | NP_001091983.1 | 172 | 106 | ||
XM_028841805.1 | XP_028697638.1 | 1604 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |