Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC42SE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257482.1 | NP_001244411.1 | 555 | 64 | ||
XM_015150416.2 | XP_015005902.1 | 802 | 64 | |||
XM_015150422.2 | XP_015005908.1 | 797 | 64 | |||
GenBank mRNA | JU335515.1 | - | 555 | 64 | ||
JU476455.1 | - | 576 | 64 | |||
JV044237.1 | - | 567 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |