Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266674.1 | NP_001253603.1 | 649 | 64 | ||
XM_015150824.2 | XP_015006310.1 | 582 | 64 | |||
XM_028828993.1 | XP_028684826.1 | 1157 | 64 | |||
GenBank mRNA | JV667309.1 | - | 649 | 64 | ||
JV669615.1 | - | 649 | 64 |
Gene Symbol: ENO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU340132.1 | - | 351 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |