Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110777.2 | NP_001104247.2 | 1244 | 61 | ||
XM_028840467.1 | XP_028696300.1 | 1130 | 61 | |||
XM_028840468.1 | XP_028696301.1 | 1660 | 61 | |||
GenBank mRNA | JU485307.1 | - | 592 | 61 | ||
JV614792.1 | - | 636 | 61 | |||
JV649669.1 | - | 312 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |