Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114353.1 | NP_001107825.1 | 322 | 99 | ||
NM_001114354.1 | NP_001107826.1 | 322 | 99 | |||
XM_015124865.2 | XP_014980351.2 | 418 | 99 | |||
XM_028838354.1 | XP_028694187.1 | 418 | 99 | |||
XM_028838355.1 | XP_028694188.1 | 418 | 99 | |||
XM_028838356.1 | XP_028694189.1 | 418 | 99 | |||
XM_028838357.1 | XP_028694190.1 | 418 | 99 | |||
XM_028838358.1 | XP_028694191.1 | 416 | 99 | |||
XM_028838359.1 | XP_028694192.1 | 416 | 99 | |||
GenBank mRNA | EU344969.1 | - | 322 | 99 | ||
EU344970.1 | - | 322 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |