Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114948.1 | NP_001108420.1 | 728 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |