Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSORS1C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114954.1 | NP_001108426.1 | 53 | 74 | ||
XM_028846821.1 | XP_028702654.1 | 1181 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |