Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115126.1 | NP_001108598.1 | 53 | 100 | ||
GenBank mRNA | EU437443.1 | - | 53 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |