Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168687.1 | NP_001162158.1 | 895 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |