Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AASDHPPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171189.1 | NP_001164660.1 | 769 | 67 | ||
GenBank mRNA | JU472738.1 | - | 845 | 67 | ||
JV043669.1 | - | 837 | 67 | |||
JV528518.1 | - | 843 | 67 | |||
JV566462.1 | - | 827 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |