Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAAR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171574.1 | NP_001165045.1 | 848 | 76 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Recommended Assay Macaca mulatta |