Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171840.1 | NP_001165311.1 | 106 | 71 | ||
XM_028834337.1 | XP_028690170.1 | 373 | 71 | |||
GenBank mRNA | AF218048.1 | - | 106 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |