Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELSPBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171843.1 | NP_001165314.1 | 635 | 77 | ||
GenBank mRNA | AF218044.1 | - | 635 | 77 |