Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFSF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205135.3 | NP_001192064.1 | 378 | 70 | ||
GenBank mRNA | JU336906.1 | - | 373 | 70 | ||
JV601371.1 | - | 357 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |