Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GXYLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193857.2 | NP_001180786.2 | 1400 | 91 | ||
XM_015151455.2 | XP_015006941.2 | 1180 | 91 | |||
GenBank mRNA | JU326216.1 | - | 971 | 91 | ||
JV648758.1 | - | 981 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |