Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TENM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193929.2 | NP_001180858.2 | 2467 | 70 | ||
XM_028848058.1 | XP_028703891.1 | 2769 | 70 | |||
XM_028848059.1 | XP_028703892.1 | 2769 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |