Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193971.2 | NP_001180900.1 | 554 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |