Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194065.1 | NP_001180994.1 | 320 | 91 | ||
GenBank mRNA | JU332579.1 | - | 155 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |