Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HS3ST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194098.3 | NP_001181027.1 | 130 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |