Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC48A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194110.2 | NP_001181039.1 | 417 | 69 | ||
XM_015151475.2 | XP_015006961.1 | 502 | 69 | |||
XM_015151476.2 | XP_015006962.1 | 464 | 69 | |||
XM_028828866.1 | XP_028684699.1 | 817 | 69 | |||
GenBank mRNA | JU335377.1 | - | 361 | 69 | ||
JU335378.1 | - | 361 | 69 | |||
JU335379.1 | - | 361 | 69 | |||
JU335380.1 | - | 361 | 69 | |||
JU335381.1 | - | 361 | 69 | |||
JU335382.1 | - | 361 | 69 | |||
JU473585.1 | - | 387 | 69 | |||
JV601998.1 | - | 420 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |