Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR8I2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194268.2 | NP_001181197.2 | 42 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |