Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX4I2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194480.2 | NP_001181409.1 | 290 | 90 | ||
XM_028827357.1 | XP_028683190.1 | 264 | 90 | |||
GenBank mRNA | JU323570.1 | - | 279 | 90 | ||
JV635837.1 | - | 273 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |