Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR10A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194487.2 | NP_001181416.2 | 652 | 99 | ||
XM_028828894.1 | XP_028684727.1 | 2099 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |